missing translation for 'onlineSavingsMsg'
Learn More

'Alginic Acid, Sodium Salt, Low Viscosity, MP Biomedicals

Product Code. 11404761 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404761 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11404761 Supplier MP Biomedicals Supplier No. 0215472580
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Source: kelp

Specifications

CAS 9005-38-3
Color White to Cream
Physical Form Powder
Quantity 100 g
Assay Percent Range >98%
Molecular Formula (C6H7O7)A(C6H7O7)BNa
Linear Formula (C6H7O6Na)n
Synonym Sodium alginate
Chemical Name or Material Alginic Acid, Sodium Salt, Low Viscosity, Viscosity (1% aq. solution): <300cps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.