missing translation for 'onlineSavingsMsg'
Learn More

Alginic acid sodium salt, low viscosity

Product Code. 11410828
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410828 100 g 100g
11420828 500 g 500g
11430828 2.5 kg 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410828 Supplier Thermo Scientific Alfa Aesar Supplier No. B25266.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa

A gelling polysaccharide extracted from giant brown seaweed, it is useful in viscosity studiesAlginic acid sodium salt is widely utilized in the food industry as flavorless gum which enhances viscosity and acts as an emulsifier. It finds application as a thickener for reactive dyestuffs which is used in textile screen printing and carpet jet-printing. It is used as indigestion tablets and for the preparation of dental impressions. As a polymer, it combines with chitosan to synthesize a biodegradable porous scaffold which is useful for tissue engineering applications. It acts as a food additive and is used in production of gel-like foods such as baking. It is also used for encapsulating agents of microparticles of beta-galactosidae and in the preparation of alginate hydrogels with tunable mechanical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
MDL Number MFCD00081310
Synonym Algin; Polymannuronic acid sodium salt

Specifications

Merck Index 14,241
Solubility Information Soluble in water. Insoluble in alcohol,chloroform and ether.
Quantity 100 g
Sensitivity Hygroscopic
Chemical Name or Material Alginic acid sodium salt, low viscosity

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.