missing translation for 'onlineSavingsMsg'
Learn More

Alginic acid sodium salt, very low viscosity

Product Code. 11418497
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418497 100 g 100g
11428497 500 g 500g
11438497 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418497 Supplier Thermo Scientific Alfa Aesar Supplier No. A18565.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa

It is a hematologic agent, drug that acts on blood and blood-forming organs and those that affect the hemostatic system. sodium alginate has been used in warp sizing, packing and printing. In printing, it is an ideal kind of material with wide applications in the cotton, wool, silk and synthetic fiber. Sodium alginate is widely used in food industry such as it is the main material of seaweed, synthetic grape and synthetic cherry, it is taken as a stabilizer of ice cream and ice cream bar because of its dense organization and slow solution speed. It is also the main material of the dainty cold powder and jelly, As a stabilizer of cake (such as cookies, bread, fine dried noodle, chocolates) and a polish of bread, sodium alginate can keep the cookies sweet-smelling and avoid falling to pieces, and make the noodle smooth and reduce breaking ratio of noodle and fragmentation of bread. Sodium alginate is also a good thickener of jam, chili sauce, jelly, tomato ketchup, fishpaste, pudding and salad flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
MDL Number MFCD00081310
Synonym Algin; Polymannuronic acid sodium salt

Specifications

Quantity 100 g
Sensitivity Hygroscopic
Merck Index 14,241
Solubility Information Soluble in water.
Chemical Name or Material Alginic acid sodium salt, very low viscosity

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.