missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ aLICator LIC Cloning and Expression Set 2 (All-in-One/WQ)

Product Code. 13263149 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
30 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13263149 30 Reactions 30 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13263149 Supplier Thermo Scientific™ Supplier No. K1291
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Get fast and efficient ligation-independent cloning and tight regulation of gene expression in E. coli. with this cloning and expression system.

aLICator LIC Cloning and Expression System is designed for fast and efficient ligation independent cloning and tight regulation of gene expression in E. coli. The pLATE bacterial expression vectors are designed for high levels of target protein expression in concert with minimal background (uninduced) expression, which permits expression of proteins that are toxic to E. coli cells. To streamline and facilitate the process of insert cloning into the expression vector, the aLICator system uses directional LIC cloning technology, a rapid procedure that provides high-cloning efficiencies.

The tightly regulated expression and fast, efficient directional cloning makes the aLICator LIC Cloning and Expression System the best choice for routine and toxic gene cloning and expression in E. coli.

Highlights

  • High efficiency LIC cloning
  • Tight control of gene expression
  • High yield expression
  • Versatile -tagged or untaged protein expression with tag removal option
TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Resistance Bacterial Ampicillin (AmpR)
Cloning Method LIC Cloning
Sample Type PCR Products
For Use With (Application) PCR Cloning
No. of Reactions 30 Reactions
Product Line aLICator
Product Type Cloning and Expression Set
Promoter T7
Protein Tag His Tag (6x)
Quantity 30 Reactions
Vector pLATE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.