missing translation for 'onlineSavingsMsg'
Learn More

Aliquat™ 336, average MW 442, Thermo Scientific™

Product Code. 10532601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10532601 250 mL Glass bottle 250mL
10398470 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10532601 Supplier Thermo Scientific Acros Supplier No. 194972500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63393-96-4
Molecular Formula C25H54ClN
Molecular Weight (g/mol) 404.16
MDL Number MFCD00011862
InChI Key XKBGEWXEAPTVCK-UHFFFAOYSA-M
Synonym methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride
PubChem CID 21218
ChEBI CHEBI:75286
IUPAC Name methyltrioctylazanium chloride
SMILES [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

Specifications

Color Amber
Density 0.8800g/mL
Flash Point 132°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4655 to 1.4715
Quantity 250 mL
Fieser 05,460; 06,404; 07,380; 13,192
Formula Weight 442
Percent Purity 88.2 to 90.6%, 112.0 to 115.0 (quaternary value)
Physical Form Viscous Liquid
Chemical Name or Material Aliquat™ 336, Average M.W. 442
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Aliquat]TS 336
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.