missing translation for 'onlineSavingsMsg'
Learn More

Alisol A 24-acetate, MedChemExpress

Product Code. 16491538
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491538 5 mg 5mg
16439138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491538 Supplier Medchem Express Supplier No. HYN0853A
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Alisol A 24-acetate (Alisol A 24-monoacetate) is a natural product.

Alisol A 24-acetate (Alisol A 24-monoacetate) is a natural product.

Specifications

Chemical Name or Material Alisol A 24-acetate
CAS 18674-16-3
Color White
Purity Grade Notes Research
Molecular Formula C32H52O6
Quantity 5 mg
Synonym Alisol A 24-monoacetate Alisol A monoacetate
Solubility Information DMSO : ≥ 50 mg/mL (93.85 mM)
SMILES C[C@]([C@@]1(C2=C([C@H](C)C[C@H](O)[C@H](C(C)(O)C)OC(C)=O)CC1)C)(CC[C@@]3([H])C4(C)C)[C@]([C@H](C2)O)([H])[C@]3(CCC4=O)C
Molecular Weight (g/mol) 532.75
Formula Weight 532.75
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.