missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alizarin, 97%, pure

Product Code. 13418388 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 13418388

Brand: Thermo Scientific Chemicals 153690025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72-48-0 | C14H8O4 | 240.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-48-0
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001201
InChI Key RGCKGOZRHPZPFP-UHFFFAOYSA-N
Synonym 1, 2-Dihydroxyanthraquinone, Mordant Red 11
PubChem CID 6293
ChEBI CHEBI:16866
IUPAC Name 1,2-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O

Specifications

Melting Point 287.0°C to 289.0°C
Boiling Point 430.0°C
CAS Min % 96.0
CAS Max % 100.0
Dye Content 96.0% min.
Assay Percent Range 97%
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in aromatic solvents, ether and hot, methanol, moderately soluble in ethanol
Infrared Spectrum Authentic
Formula Weight 240.21
Loss on Drying 2% max.
Percent Purity 97%
Grade Pure
Packaging Plastic drum
Quantity 2.5 kg
Physical Form Fine Powder
Chemical Name or Material Alizarin, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Alizarin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.