missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S (Sodium Sulfonate), MP Biomedicals™

Product Code. 11443060 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11443060 25 g 25g
11453060 100 g 100g
11463060 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11443060

Brand: MP Biomedicals 0210037525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
MDL Number MFCD00013049
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
Synonym Sodium Alizarinsulfonate, Alizarin sodium sulfonate
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

Formula Weight 342.3
Color Orange
Quantity 25 g
Physical Form Powder
Chemical Name or Material Alizarin Red S, Indicator: pH Range 3.7 - 5.2
compliance-icons
Product Identifier
  • Alizarin Red S
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Alizarin Red S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.