missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alizarin Yellow R sodium salt, 96%, pure

Product Code. 10711632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10711632 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10711632 Supplier Thermo Scientific Chemicals Supplier No. 400490250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1718-34-9 | C13H8N3NaO5 | 309.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1718-34-9
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C13H8N3NaO5
MDL Number MFCD00067121
Synonym 5-(4-Nitrophenylazo)-salicylic acid sodium salt, Mordant Yellow 3R, 2-Hydroxy-5-((4-nitrophenyl)azo)benzoic acid monosodium salt
InChI Key HKSANXZSKDUDBD-UHFFFAOYSA-M
Infrared Spectrum Authentic
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]
IUPAC Name sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular Weight (g/mol) 309.213
PubChem CID 44135758
Formula Weight 309.2
Loss on Drying 5% max. (105°C)
Grade Pure
Packaging Glass bottle
UV_Vis 95% min.
Visual Transition Interval from pH 10.2 (yellow) to pH 12.0 (red)
Color Brown to Orange
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Alizarin Yellow R sodium salt, 96%
Show More Show Less
compliance-icons
Product Identifier
  • Alizarin Yellow R sodium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.