missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Yellow R, Sodium Salt, TRC

Product Code. 30179511
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179511 5 g 5g
30179272 1 g 1g
30179481 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179511 Supplier Toronto Research Chemicals Supplier No. TRCA3585055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1718-34-9
Formula Weight 309.0362
InChI Formula InChI=1S/C13H9N3O5.Na/c17-12-6-3-9(7-11(12)13(18)19)15-14-8-1-4-10(5-2-8)16(20)21;/h1-7,17H,(H,18,19);/q;+1/p-1
IUPAC Name sodium;2-hydroxy-5-[(E)-(4-nitrophenyl)diazenyl]benzoate
Molecular Formula C13H8N3O5Na
Molecular Weight (g/mol) 309.21
SMILES [Na+].Oc1ccc(cc1C(=O)[O-])N=Nc2ccc(cc2)[N+](=O)[O-]
Chemical Name or Material Alizarin Yellow R, Sodium Salt

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.