missing translation for 'onlineSavingsMsg'
Learn More

Alizarine S solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12947153 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12947153 1000 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12947153 Supplier Fisher Chemical Supplier No. RP 006 100
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 121-17-5
Concentration 1.0N
Molecular Formula C7H3ClF3NO2
MDL Number MFCD00007084
InChI Key TZGFQIXRVUHDLE-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
IUPAC Name 1-chloro-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol) 225.55
PubChem CID 8462
Grade Eur Ph Ref.: 1002601
Packaging Bottle
Quantity 1000 mL
Chemical Name or Material Aminohippuric acid reagent.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.