missing translation for 'onlineSavingsMsg'
Learn More

Alkali Blue 6B Solution, Solstice

Product Code. 15688360
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15688360 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15688360 Supplier Honeywell-Fluka Supplier No. 32806250ML
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket
This item is not returnable. View return policy

0.1% in ethanol, for acid-base titration

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 1324-80-7
Molecular Formula C37H30N3NaO4S
UN Number 1170
InChI Key BPCZPTGXEYXQRS-UHFFFAOYSA-M
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)C(C3=CC=C(C=C3)NC4=CC=CC=C4)(C5=CC=C(C=C5)NC6=CC=CC=C6S(=O)(=O)[O-])O.[Na+]
IUPAC Name sodium;2-[4-[bis(4-anilinophenyl)-hydroxymethyl]anilino]benzenesulfonate
Molecular Weight (g/mol) 635.714
PubChem CID 71464047
Formula Weight 635.71g/mol
Quantity 250 mL
Flash Point 14°C (57.2°F)
Chemical Name or Material Alkali Blue 6B Solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Alkali blue 6B solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, C.I. Pigment blue 61
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.