missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase Mouse, Clone: ALPL/597, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 249-MSM1-P0
Additional Details : Weight : 0.01000kg
Specifications
Alkaline Phosphatase | |
Cell Surface | |
ALPL/597 | |
Unconjugated | |
NP_016856392 | |
Mouse | |
55kDa | |
Protein A/G Purified | |
RUO | |
249 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
ALPL | |
Recombinant human ALPL protein | |
Intestine | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |