missing translation for 'onlineSavingsMsg'
Learn More

all-trans-Canthaxanthin, TRC

Product Code. 30010034
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010034 25 mg 25mg
30012154 5 mg 5mg
30008565 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010034 Supplier Toronto Research Chemicals Supplier No. TRCC17573525MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 514-78-3
Molecular Formula C40 H52 O2
Molecular Weight (g/mol) 564.84
Synonym β-Carotene-4,4'-dione (6CI), β-Carotene-4,4'-dione, all-trans- (8CI), 4,4'-Diketo-β-carotene, 4,4'-Dioxo-β-carotene, C.I. 40850, C.I. Food Orange 8, Canthaxanthin, Canthaxanthine, Carophyll red, E 161g, Food Orange 8, Lucantin Red, NSC 374110, Roxanthin Red 10, all-trans-Canthaxanthin
IUPAC Name 2,4,4-trimethyl-3-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyl-18-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)octadeca-1,3,5,7,9,11,13,15,17-nonaenyl]cyclohex-2-en-1-one
SMILES C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C(=O)CCC1(C)C)\C=C\C=C(/C)\C=C\C2=C(C)C(=O)CCC2(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C40H52O2/c1-29(17-13-19-31(3)21-23-35-33(5)37(41)25-27-39(35,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-36-34(6)38(42)26-28-40(36,9)10/h11-24H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+
Formula Weight 564.3967
Percent Purity >95%
Chemical Name or Material Canthaxanthin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.