missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies All-Trans Retinoic Acid

Product Code. p-7217364
Encompass_Preferred
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
This item is not returnable. View return policy

Product Code. 15855521

Brand: STEMCELL Technologies 72262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Retinoid pathway activator; Activates retinoic acid receptor (RAR)

  • All-Trans Retinoic Acid is a derivative of Vitamin A that functions as a ligand for the retinoic acid receptor (RAR, IC50 = 14 nM)
  • RARs heterodimerize with retinoid X receptors (RXRs) and bind to retinoic acid response elements (RAREs) in DNA and act as transcription factors, altering gene expression
  • (Apfel et al.), Chambon)

Specifications

CAS 302-79-4
Synonym ATRA; NSC 122758; Retinoic acid; Trans retinoic acid; Tretinoin; Vitamin A acid
Quantity 50 mg
Molecular Formula C20H28O2
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.