missing translation for 'onlineSavingsMsg'
Learn More

all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized, Thermo Scientific Chemicals

Product Code. 15255988 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15255988 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15255988 Supplier Thermo Scientific Acros Supplier No. 346325000
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-47-9
InChI Key QGNJRVVDBSJHIZ-QHLGVNSISA-N
Synonym retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
PubChem CID 638034
ChEBI CHEBI:32095
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C

Specifications

Flash Point >100°C
Molecular Formula C22H32O2
Refractive Index 1.5470 to 1.5550
Quantity 500 g
Merck Index 15,10211
Solubility Information Solubility in water: difficult soluble.
Molecular Weight (g/mol) 328.49
Viscosity 20 mPa.s (60°C)
Formula Weight 328.49
Percent Purity 1.50 to 1.65 million IU/g (Vitamin A) Photometr.
Related Substances 0.14 (A370/A326) max
Chemical Name or Material all-trans-Retinyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.