missing translation for 'onlineSavingsMsg'
Learn More

Brady™ All-Weather Vinyl Labels

Product Code. 31910400 Shop All Brady Products
Change view
Click to view available options
Color:
Black/Orange on White
Black/Yellow on White
Blue on White
Red on White
Unit Size:
100 rolls
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31910400 Red on White 100 rolls
31910386 Black/Yellow on White 100 rolls
31910378 Black/Orange on White 100 rolls
31910395 Blue on White 100 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31910400 Supplier Brady™ Supplier No. 177769
  • ONLINE EXCLUSIVE PRICE
    81.15 €
    78.98 € / 100 rolls
    Save 2.17 €
    2% Off
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This durable, low-shrink vinyl with ultra-aggressive adhesive is ideal for labelling pipes, equipment, shelves and nearly any other industrial surface.

  • Pre-printed Spanish headers allow you to quickly create multicolor signs with a single print pass
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces - including outdoors
  • Conforms and adheres to difficult surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to chemicals, greases and oils, salt spray, cleaners, detergents and general industrial grime
  • A ribbon is required to print this material - use part number B30-R10000

Specifications

Certifications/Compliance BS 5609:1986 Section 2 and Section 3
Adhesive Type Permanent Acrylic
Color Red on White
Finish Gloss
Format Die-cut
Material Vinyl
Print Technology Thermal Transfer
Product Type Label
For Use With (Application) Electrical Labeling, General Identification, Hazard Communication Identification, Pipe and Valve Marking, Safety Identification
For Use With (Equipment) M610, M611, M710, BMP61, BMP71
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Shape Rectangle
Temperature Range (English) -40°F to +180°F
Temperature Range (Metric) -40°C to +82°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Length (English) 16 ft.
Width (English) 4 in.
Width (Metric) 101.6 mm
Width (English) Printable Area 101.6 mm
Width (Metric) Printable Area 101.6 mm
Height (English) 1.9 in.
Height (English) Printable Area 1.4 in.
Height (Metric) 48.26 mm
Height (Metric) Printable Area 35.56 mm
Alternate Part Number M6-38-595-PEL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.