missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, 98%

Product Code. 10142320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10142320 500 g 500g
10745741 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10142320 Supplier Thermo Scientific Acros Supplier No. 102845000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-59-6 | C4H6N4O3 | 158.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-59-6
Molecular Formula C4H6N4O3
Molecular Weight (g/mol) 158.12
MDL Number MFCD00005260
InChI Key POJWUDADGALRAB-UHFFFAOYSA-N
Synonym allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream
PubChem CID 204
ChEBI CHEBI:15676
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
SMILES C1(C(=O)NC(=O)N1)NC(=O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Beilstein 25, V,15, 338
Merck Index 15, 250
Solubility Information Solubility in water: 1g/190mL. Other solubilities: readily soluble in alkalies,1g/500mL alcohol,practically insoluble in ether
Formula Weight 158.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Allantoin, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.