missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, TRC

Product Code. 30132784
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132784 25 g 25g
30138275 5 g 5g
30011897 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132784 Supplier Toronto Research Chemicals Supplier No. TRCA54050025G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-59-6
Molecular Formula C4 H6 N4 O3
Molecular Weight (g/mol) 158.12
Synonym Allantoin, (RS)-(2,5-dioxoimidazolidin-4-yl)urea, Urea, N-(2,5-dioxo-4-imidazolidinyl)-Allantoin (8CI), Urea, (2,5-dioxo-4-imidazolidinyl)- (9CI), (2,5-Dioxo-4-imidazolidinyl)urea, (±)-Allantoin, 5-Ureidohydantoin, Allantion, Allantol, Cordianine, DL-Allantoin, Glyoxyldiureid, Glyoxyldiureide, Glyoxylic diureide, NSC 7606, Psoralon, Sebical, Septalan
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
SMILES NC(=O)NC1NC(=O)NC1=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H6N4O3/c5-3(10)6-1-2(9)8-4(11)7-1/h1H,(H3,5,6,10)(H2,7,8,9,11)
Formula Weight 158.044
Percent Purity >95%
Chemical Name or Material Allantoin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.