missing translation for 'onlineSavingsMsg'
Learn More

Allenylboronic acid pinacol ester, TRC

Product Code. 30324522
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324522 100 mg 100mg
30324457 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30324522 Supplier Toronto Research Chemicals Supplier No. TRCA542003100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allenylboronic acid pinacol ester
CAS 865350-17-0
Molecular Formula C9H15BO2
Quantity 100 mg
InChI Formula InChI=1S/C9H15BO2/c1-6-7-10-11-8(2,3)9(4,5)12-10/h7H,1H2,2-5H3
SMILES CC1(C)OB(OC1(C)C)C=C=C
IUPAC Name 4,4,5,5-tetramethyl-2-propa-1,2-dienyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 166.03
Formula Weight 166.1165
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.