missing translation for 'onlineSavingsMsg'
Learn More

Allopurinol Inhibitor of Xanthine Oxidase MP Biomedicals

Product Code. 11474592 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474592 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474592 Supplier MP Biomedicals Supplier No. 0219015005
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 315-30-0
Molecular Formula C5H4N4O
MDL Number MFCD00599413
Quantity 5 g
Synonym allopurinol, 1h-pyrazolo 3,4-d pyrimidin-4-ol, zyloprim, zyloric, lopurin, atisuril, bleminol, caplenal, uripurinol, embarin
InChI Key OFCNXPDARWKPPY-UHFFFAOYSA-N
SMILES O=C1N=CN=C2NNC=C12
IUPAC Name 1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
Molecular Weight (g/mol) 136.11
PubChem CID 2094
ChEBI CHEBI:40279
Formula Weight 136.1
Chemical Name or Material Allopurinol, Inhibitor of Xanthine Oxidase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.