missing translation for 'onlineSavingsMsg'
Learn More

Manutan Alloy rope hook with catch, medium duty, 10mm diameter

Product Code. 12871379
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12871379 Supplier Manutan Supplier No. A339948
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Complete with catch. Available with standard or with swivel hook.

  • Standard
  • Rope diameter: 10mm
  • WLL: 1000kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.