missing translation for 'onlineSavingsMsg'
Learn More

Manutan Alloy swivel rope hook with catch, medium duty, 13mm diameter

Product Code. 12871399
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12871399 Supplier Manutan Supplier No. A339943
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Complete with catch. Available with standard or with swivel hook.

  • Swivel
  • Rope diameter: 13mm
  • WLL: 2000kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.