missing translation for 'onlineSavingsMsg'
Learn More

Allyl acetate, 97%

Product Code. 11304068
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304068 50 mL 50mL
11394058 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304068 Supplier Thermo Scientific Alfa Aesar Supplier No. 041353.AD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-87-7 | C5H8O2 | 100.117 g/mol

Ally acetate is used in industrial intermediate. Used in synthesis of colophony and bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-87-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00008721
InChI Key FWZUNOYOVVKUNF-UHFFFAOYSA-N
Synonym allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i
PubChem CID 11584
IUPAC Name prop-2-enyl acetate
SMILES CC(=O)OCC=C

Specifications

Melting Point 6°C
Density 0.928 g/mL
Boiling Point 103°C to 104°C
Refractive Index 1.4042
Quantity 50 mL
UN Number UN2333
Merck Index 14,285
Solubility Information Soluble in ethanol. Miscible with ethanol and ether. Slightly soluble in water.
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Allyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.