missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloride, 98%, stab. with propylene oxide, Thermo Scientific Chemicals

Product Code. 11466887
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466887 2.5 L 2500mL
11456887 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466887 Supplier Thermo Scientific Alfa Aesar Supplier No. A14330.0F
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-05-1 | Molecular Formula: C3H5Cl | Molecular Weight: 76.523 g/mol

Allyl chloride is used as an alkylating agent. It is used in treatment with sodium bis(trimethylsilyl)amide gives the strained, highly reactive alkene cyclopropene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl chloride is used as an alkylating agent. It is used in treatment with sodium bis(trimethylsilyl)amide gives the strained, highly reactive alkene cyclopropene.

Solubility
Soluble in water (3600 mg/L at 20°C), ethanol, ether, petroleum, acetone, chloroform, benzene, methanol and 2-prop.

Notes
Store at -20°C. Keep tightly closed in a dry and cool place. Keep in properly labeled containers. Keep containers tightly closed in a cool, well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents, boron trifluoride, sulfuric acid and nitric acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-05-1
Molecular Formula C3H5Cl
MDL Number MFCD00000984
InChI Key OSDWBNJEKMUWAV-UHFFFAOYSA-N
Synonym allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
PubChem CID 7850
ChEBI CHEBI:82379
IUPAC Name 3-chloroprop-1-ene

Specifications

Melting Point -135°C
Density 0.939
Boiling Point 44°C to 46°C
Flash Point -28°C
Odor Sweet
Linear Formula CH2=CHCH2Cl
Refractive Index 1.4155
Quantity 2.5 L
UN Number UN1100
Beilstein 635704
Merck Index 14,289
Solubility Information Soluble aq. soln. (3600 mg/L (at 20°C),ethanol,ether,petroleum,acetone,chloroform,benzene,methanol and 2-prop.)
SMILES ClCC=C
Molecular Weight (g/mol) 76.52
Formula Weight 76.53
Percent Purity 98%
Chemical Name or Material Allyl chloride, stabilized with propylene oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.