missing translation for 'onlineSavingsMsg'
Learn More

Allyl Chloride, TRC

Product Code. 30324998
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324998 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324998 Supplier Toronto Research Chemicals Supplier No. TRCA54971550ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allylchloride
CAS 107-05-1
Molecular Formula C3 H5 Cl
Quantity 50 mL
Synonym Propene, 3-chloro- (8CI), 3-Chloro-1-propene, 1-Chloro-2-propene, 2-Propenyl chloride, 3-Chloropropene, 3-Chloropropylene, Allyl chloride, HCO 1260ZF, NSC 20939
InChI Formula InChI=1S/C3H5Cl/c1-2-3-4/h2H,1,3H2
SMILES ClCC=C
IUPAC Name 3-chloroprop-1-ene
Molecular Weight (g/mol) 76.52
Formula Weight 76.008
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.