missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloroacetate, 98%, Thermo Scientific™

Product Code. 10554711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass Bottle
Unit Size:
5mL
Product Code. Quantity Packaging unitSize
10554711 5 mL Glass Bottle 5mL
1 options
This item is not returnable. View return policy

Product Code. 10554711

Brand: Thermo Scientific Acros 335150050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2916-14-5
Molecular Formula C5H7ClO2
Molecular Weight (g/mol) 134.56
MDL Number MFCD00058936
InChI Key VMBJJCDVORDOCF-UHFFFAOYSA-N
Synonym allyl chloroacetate, allyl 2-chloroacetate, chloroacetic acid allyl ester, acetic acid, chloro-, 2-propenyl ester, prop-2-en-1-yl 2-chloroacetate, acetic acid, chloro-, allyl ester, allyl monochloroacetate, acmc-1cffa, chloroacetic acid,2-propenyl ester
PubChem CID 76206
IUPAC Name prop-2-enyl 2-chloroacetate
SMILES C=CCOC(=O)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1590g/mL
Boiling Point 164°C (265.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4450 to 1.447
Quantity 5 mL
Specific Gravity 1.159
Formula Weight 134.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allyl chloroacetate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Allyl chloroacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.