missing translation for 'onlineSavingsMsg'
Learn More

Allyl cyanide, 97%

Product Code. 11480031
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480031 25 g 25g
11490031 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480031 Supplier Thermo Scientific Alfa Aesar Supplier No. 044775.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-75-1 | C4H5N | 67.091 g/mol

Allyl cyanide is used as an anti-microbial. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (3 g/L) at 20°C.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-75-1
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
MDL Number MFCD00001962
InChI Key SJNALLRHIVGIBI-UHFFFAOYSA-N
Synonym 3-butenenitrile, allyl cyanide, 3-butenylnitrile, 3-butenonitrile, allylnitrile, vinylacetonitrile, 3-cyanopropene, 3-cyano-1-propene, 1-butene-4-nitrile, propene-3-nitrile
PubChem CID 8009
IUPAC Name but-3-enenitrile
SMILES C=CCC#N

Specifications

Melting Point -87°C
Color Brown to Colorless
Density 0.834 g/mL
Boiling Point 118°C to 120°C
Flash Point 24°C (75°F)
Odor Unpleasant
Linear Formula CH2=CHCH2CN
Refractive Index 1.405
Quantity 25 g
UN Number UN1992
Beilstein 605352
Merck Index 14,290
Solubility Information Not miscible in water.
Formula Weight 67.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allyl cyanide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.