Learn More
Allyl cyanide, 97%
Description
Allyl cyanide is used as an anti-microbial. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -87°C |
| Color | Brown to Colorless |
| Density | 0.834 g/mL |
| Boiling Point | 118°C to 120°C |
| Flash Point | 24°C (75°F) |
| Odor | Unpleasant |
| Linear Formula | CH2=CHCH2CN |
| Refractive Index | 1.405 |
| Quantity | 100 g |
| UN Number | UN1992 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.