missing translation for 'onlineSavingsMsg'
Learn More

Allyl Glycidyl Ether, 99+%

Product Code. 10598650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10598650 500 g Glass bottle 500g
10356210 100 g Glass bottle 100g
10761165 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10598650 Supplier Thermo Scientific Acros Supplier No. 102945000
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-92-3 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless
Density 0.9620g/mL
Boiling Point 154.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4332 to 1.4352
Quantity 500 g
Specific Gravity 0.962
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether and,benzene
Formula Weight 114.14
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Allyl glycidyl ether, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl glycidyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361f-Suspected of damaging fertility.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.