missing translation for 'onlineSavingsMsg'
Learn More

Allyl heptanoate, 97%, Thermo Scientific™

Product Code. 11312936
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312936 500 g 500g
11302936 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312936 Supplier Thermo Scientific Alfa Aesar Supplier No. L10671.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-19-8
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00038341
InChI Key SJWKGDGUQTWDRV-UHFFFAOYSA-N
Synonym allyl heptanoate, allyl heptylate, 2-propenyl heptanoate, allyl enanthate, allyl heptoate, heptanoic acid, 2-propenyl ester, prop-2-en-1-yl heptanoate, heptanoic acid, allyl ester, allylester kyseliny enanthove, heptanoic acid, 2-propen-1-yl ester
PubChem CID 8878
IUPAC Name prop-2-enyl heptanoate
SMILES CCCCCCC(=O)OCC=C

Specifications

Density 0.882
Boiling Point 210°C to 212°C
Flash Point 74°C (165°F)
Linear Formula CH3(CH2)5CO2CH2CH=CH2
Refractive Index 1.4291
Quantity 500 g
UN Number UN2810
Beilstein 8544440
Formula Weight 170.25
Percent Purity 97%
Chemical Name or Material Allyl heptanoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.