missing translation for 'onlineSavingsMsg'
Learn More

Allyl iodide, 97%, stabilized

Product Code. 10236370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10236370 25 mL Glass bottle 25mL
10216420 100 mL Glass bottle 100mL
11954501 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10236370 Supplier Thermo Scientific Acros Supplier No. 218540250
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 556-56-9 | C3H5I | 167.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-56-9
Molecular Formula C3H5I
Molecular Weight (g/mol) 167.98
MDL Number MFCD00001094
InChI Key HFEHLDPGIKPNKL-UHFFFAOYSA-N
Synonym allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
PubChem CID 11166
IUPAC Name 3-iodoprop-1-ene
SMILES ICC=C

Specifications

Melting Point -99.0°C
Color Brown-Red or Orange-Red to Yellow
Density 1.8300g/mL
Boiling Point 102.0°C to 103.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2I
Refractive Index 1.5530 to 1.5550
Quantity 25 mL
Beilstein 01,202
Merck Index 15,285
Specific Gravity 1.83
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
Stabilizer Copper
Formula Weight 167.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allyl iodide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl iodide, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.