missing translation for 'onlineSavingsMsg'
Learn More

Allyl isothiocyanate, 94%, stab.

Product Code. 11406720
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406720 250 g 250g
11486710 5 g 5g
11496710 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406720 Supplier Thermo Scientific Alfa Aesar Supplier No. L02901.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57-06-7 | C4H5NS | 99.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-06-7
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.151
MDL Number MFCD00004822
InChI Key ZOJBYZNEUISWFT-UHFFFAOYSA-N
Synonym allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
PubChem CID 5971
ChEBI CHEBI:73224
IUPAC Name 3-isothiocyanatoprop-1-ene
SMILES C=CCN=C=S

Specifications

Melting Point -80°C
Density 1.013
Boiling Point 149°C to 151°C
Flash Point 46°C (114°F)
Odor Pungent
Refractive Index 1.53
Quantity 250 g
UN Number UN1545
Beilstein 773748
Sensitivity Moisture sensitive
Merck Index 14,295
Formula Weight 99.16
Percent Purity 94%
Chemical Name or Material Allyl isothiocyanate, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.