missing translation for 'onlineSavingsMsg'
Learn More

Allyl isothiocyanate, 94%, stab., Thermo Scientific Chemicals

Catalog Number 11486710
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy
57-06-7
C4H5NS
99.151
MFCD00004822
ZOJBYZNEUISWFT-UHFFFAOYSA-N
allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
5971
CHEBI:73224
3-isothiocyanatoprop-1-ene
C=CCN=C=S
This item is not returnable. View return policy
57-06-7
C4H5NS
99.151
MFCD00004822
ZOJBYZNEUISWFT-UHFFFAOYSA-N
allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
5971
CHEBI:73224
3-isothiocyanatoprop-1-ene
C=CCN=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -80°C
Density 1.013
Boiling Point 149°C to 151°C
Flash Point 46°C (114°F)
Odor Pungent
Refractive Index 1.53
Quantity 5 g
UN Number UN1545
Beilstein 773748
Sensitivity Moisture sensitive
Merck Index 14,295
Formula Weight 99.16
Percent Purity 94%
Chemical Name or Material Allyl isothiocyanate, stabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'