missing translation for 'onlineSavingsMsg'
Learn More

Allyl Isothiocyanate, TRC

Product Code. 30011156
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011156 250 mg 250mg
30007648 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011156 Supplier Toronto Research Chemicals Supplier No. TRCA556610250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-06-7
Molecular Formula C4 H5 N S
Molecular Weight (g/mol) 99.15
Synonym Isothiocyanic acid, allyl ester (6CI,8CI), 3-Isothiocyanato-1-propene, 2-Propenyl isothiocyanate, AITC, Allyl isothiocyanate, Allyl thioisocyanate, Allylsevenolum, Allyspol, Allyspol 75EC, Carbospol, NSC 5572, Oleum sinapis, Redskin, Senfoel, Wasaouro, Wasaouro EXT
IUPAC Name 3-isothiocyanatoprop-1-ene
SMILES C=CCN=C=S

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H5NS/c1-2-3-5-4-6/h2H,1,3H2
Formula Weight 99.0143
Chemical Name or Material Allyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.