missing translation for 'onlineSavingsMsg'
Learn More

Allyl Isothiocyanate, TRC

Product Code. 30316153
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316153 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316153 Supplier Toronto Research Chemicals Supplier No. TRCA556610100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allyl isothiocyanate
CAS 57-06-7
Molecular Formula C4 H5 N S
Quantity 100 mg
Synonym Isothiocyanic acid, allyl ester (6CI,8CI), 3-Isothiocyanato-1-propene, 2-Propenyl isothiocyanate, AITC, Allyl isothiocyanate, Allyl thioisocyanate, Allylsevenolum, Allyspol, Allyspol 75EC, Carbospol, NSC 5572, Oleum sinapis, Redskin, Senfoel, Wasaouro, Wasaouro EXT
InChI Formula InChI=1S/C4H5NS/c1-2-3-5-4-6/h2H,1,3H2
SMILES C=CCN=C=S
IUPAC Name 3-isothiocyanatoprop-1-ene
Molecular Weight (g/mol) 99.15
Formula Weight 99.0143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.