missing translation for 'onlineSavingsMsg'
Learn More

Allyl Phenyl Ether, TRC

Product Code. 30315186
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315186 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315186 Supplier Toronto Research Chemicals Supplier No. TRCA57221010G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allyl Phenyl Ether
CAS 1746-13-0
Molecular Formula C9H10O
Quantity 10 g
Synonym Allyloxybenzene, NSC 4746, Phenyl 2-Propenyl Ether, Phenyl Allyl Ether, Phenylpropenyl Ether, (2-Propenyloxy)benzene, 1-(2-Propenyloxy)benzene, 1-Phenoxy-2-propene, 3-Phenoxy-1-propene, 3-Phenoxypropene, Ether Allyl Phenyl, (2-Propenyloxy)benzene
InChI Formula InChI=1S/C9H10O/c1-2-8-10-9-6-4-3-5-7-9/h2-7H,1,8H2
SMILES C=CCOc1ccccc1
IUPAC Name prop-2-enoxybenzene
Molecular Weight (g/mol) 134.18
Formula Weight 134.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.