missing translation for 'onlineSavingsMsg'
Learn More

Allylamine, 98+%

Product Code. 11456740
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456740 100 mL 100mL
11466740 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456740 Supplier Thermo Scientific Alfa Aesar Supplier No. L07162.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 107-11-9 | C3H7N | 57.10 g/mol

Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-11-9
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.10
MDL Number MFCD00008199
InChI Key VVJKKWFAADXIJK-UHFFFAOYSA-N
Synonym allylamine, 2-propen-1-amine, monoallylamine, 3-aminopropylene, 3-aminopropene, allyl amine, 2-propenamine, 3-amino-1-propene, 2-propenylamine, polyallylamine
PubChem CID 7853
IUPAC Name prop-2-en-1-amine
SMILES NCC=C

Specifications

Melting Point -88°C
Density 0.761
Boiling Point 53°C to 54°C
Flash Point −28°C (−18°F)
Odor Ammonia-like
Linear Formula CH2=CHCH2NH2
Refractive Index 1.421
Quantity 100 mL
UN Number UN2334
Beilstein 635703
Sensitivity Air Sensitive
Merck Index 14,287
Solubility Information Fully miscible in water.
Formula Weight 57.1
Percent Purity ≥98%
Chemical Name or Material Allylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.