Learn More
Allylboronic acid pinacol ester, 98+%
Description
Allylboronic acid pinacol ester reacts with carboxylic acids, in the presence of tri-n-butyltin hydride, to give homoallylic alcohols in good yield. Homoallylic alcohols can also be formed by allylboration of aldehydes. It is a reagent used for palladium-catalyzed Suzuki-Miyaura cross-coupling reactions and olefin metathesis, intermolecular radical additions, allylboration of aldehydes catalyzed by chiral spirobiindane diol (SPINOL) based phosphoric acids, cobalt-catalyzed regioselective hydrovinylation of dienes with alkenes, nucleic acid-templated energy transfer leading to a photorelease reaction and stereoselective indium-catalyzed Hosomi-Sakurai reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Allylboronic acid pinacol ester reacts with carboxylic acids, in the presence of tri-n-butyltin hydride, to give homoallylic alcohols in good yield. Homoallylic alcohols can also be formed by allylboration of aldehydes. It is a reagent used for palladium-catalyzed Suzuki-Miyaura cross-coupling reactions and olefin metathesis, intermolecular radical additions, allylboration of aldehydes catalyzed by chiral spirobiindane diol (SPINOL) based phosphoric acids, cobalt-catalyzed regioselective hydrovinylation of dienes with alkenes, nucleic acid-templated energy transfer leading to a photorelease reaction and stereoselective indium-catalyzed Hosomi-Sakurai reactions.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.89 |
| Boiling Point | 50°C to 53°C (5 mmHg) |
| Flash Point | 46°C (140°F) |
| Refractive Index | 1.427 |
| Quantity | 1 g |
| UN Number | UN3272 |
| Beilstein | 4244068 |
| Solubility Information | Not miscible or difficult to mix in water. |
| IUPAC Name | 4,4,5,5-tetramethyl-2-prop-2-enyl-1,3,2-dioxaborolane |
| Formula Weight | 168.05 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.