missing translation for 'onlineSavingsMsg'
Learn More

Allylboronic acid pinacol ester, 98+%

Product Code. 11352947
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11352947 5 g 5g
11342947 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11352947

Brand: Thermo Scientific Alfa Aesar L16232.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72824-04-5 | C9H17BO2 | 168.04 g/mol

Allylboronic acid pinacol ester reacts with carboxylic acids, in the presence of tri-n-butyltin hydride, to give homoallylic alcohols in good yield. Homoallylic alcohols can also be formed by allylboration of aldehydes. It is a reagent used for palladium-catalyzed Suzuki-Miyaura cross-coupling reactions and olefin metathesis, intermolecular radical additions, allylboration of aldehydes catalyzed by chiral spirobiindane diol (SPINOL) based phosphoric acids, cobalt-catalyzed regioselective hydrovinylation of dienes with alkenes, nucleic acid-templated energy transfer leading to a photorelease reaction and stereoselective indium-catalyzed Hosomi-Sakurai reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylboronic acid pinacol ester reacts with carboxylic acids, in the presence of tri-n-butyltin hydride, to give homoallylic alcohols in good yield. Homoallylic alcohols can also be formed by allylboration of aldehydes. It is a reagent used for palladium-catalyzed Suzuki-Miyaura cross-coupling reactions and olefin metathesis, intermolecular radical additions, allylboration of aldehydes catalyzed by chiral spirobiindane diol (SPINOL) based phosphoric acids, cobalt-catalyzed regioselective hydrovinylation of dienes with alkenes, nucleic acid-templated energy transfer leading to a photorelease reaction and stereoselective indium-catalyzed Hosomi-Sakurai reactions.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72824-04-5
Molecular Formula C9H17BO2
Molecular Weight (g/mol) 168.04
MDL Number MFCD00013347
InChI Key YMHIEPNFCBNQQU-UHFFFAOYSA-N
Synonym allylboronic acid pinacol ester, 2-allyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, allylboronic acid, pinacol cyclic ester, 4,4,5,5-tetramethyl-2-prop-2-en-1-yl-1,3,2-dioxaborolane, allylboronic acid piracol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-propenyl, 2-prop-2-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, allylboronicacidpinacolester, pinacol allylboronate, abape
PubChem CID 2763171
SMILES CC1(C)OB(CC=C)OC1(C)C

Specifications

Density 0.89
Boiling Point 50°C to 53°C (5 mmHg)
Flash Point 46°C (140°F)
Refractive Index 1.427
Quantity 5 g
UN Number UN3272
Beilstein 4244068
Solubility Information Not miscible or difficult to mix in water.
IUPAC Name 4,4,5,5-tetramethyl-2-(prop-2-en-1-yl)-1,3,2-dioxaborolane
Formula Weight 168.05
Percent Purity ≥98%
Chemical Name or Material Allylboronic acid pinacol ester
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Allylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.