missing translation for 'onlineSavingsMsg'
Learn More

Allylboronic acid pinacol ester, 98%

Product Code. 10695145 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10695145 1 g Glass bottle 1g
10565924 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10695145 Supplier Thermo Scientific Acros Supplier No. 395130010
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 72824-04-5 | C9H17BO2 | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72824-04-5
Molecular Formula C9H17BO2
Molecular Weight (g/mol) 168.04
MDL Number MFCD00013347
InChI Key YMHIEPNFCBNQQU-UHFFFAOYSA-N
Synonym allylboronic acid pinacol ester, 2-allyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, allylboronic acid, pinacol cyclic ester, 4,4,5,5-tetramethyl-2-prop-2-en-1-yl-1,3,2-dioxaborolane, allylboronic acid piracol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-propenyl, 2-prop-2-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, allylboronicacidpinacolester, pinacol allylboronate, abape
PubChem CID 2763171
IUPAC Name 4,4,5,5-tetramethyl-2-prop-2-enyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(CC=C)OC1(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8900g/mL
Boiling Point 50.0°C to 53.0°C (5.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble
Formula Weight 168.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allylboronic acid pinacol ester
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.