missing translation for 'onlineSavingsMsg'
Learn More

Allylboronic acid pinacol ester, TRC

Product Code. 30323141
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323141 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323141 Supplier Toronto Research Chemicals Supplier No. TRCA614258500MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allylboronic acid pinacol ester
CAS 72824-04-5
Molecular Formula C9H17BO2
Quantity 500 mg
InChI Formula InChI=1S/C9H17BO2/c1-6-7-10-11-8(2,3)9(4,5)12-10/h6H,1,7H2,2-5H3
SMILES CC1(C)OB(CC=C)OC1(C)C
IUPAC Name 4,4,5,5-tetramethyl-2-prop-2-enyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 168.04
Formula Weight 168.1322
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.