missing translation for 'onlineSavingsMsg'
Learn More

Allylcyclopentane, 97%

Product Code. 11439696
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439696 1 g 1g
11449696 5 g 5g
11459696 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439696 Supplier Thermo Scientific Alfa Aesar Supplier No. B20228.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3524-75-2 | C8H14 | 110.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3524-75-2
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00001390
InChI Key NHIDGVQVYHCGEK-UHFFFAOYSA-N
Synonym allylcyclopentane, 3-cyclopentylpropene, cyclopentane, 2-propenyl, cyclopentane, allyl, 3-cyclopentyl-1-propene, allyl cyclopentane, 2-propenylcyclopentane, 1-propene, 3-cyclopentyl, 3-cyclopentylprop-1-ene
PubChem CID 77059
IUPAC Name prop-2-enylcyclopentane
SMILES C=CCC1CCCC1

Specifications

Melting Point -111°C
Density 0.793
Boiling Point 125°C to 127°C
Flash Point 12°C (53°F)
Refractive Index 1.441
Quantity 1 g
UN Number UN3295
Beilstein 2036419
Formula Weight 110.2
Percent Purity 98%
Chemical Name or Material Allylcyclopentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.