missing translation for 'onlineSavingsMsg'
Learn More

Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™

Product Code. 10735135 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10735135 800 mL AcroSeal™ Glass bottle 800mL
10081544 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10735135 Supplier Thermo Scientific Acros Supplier No. 209538000
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1730-25-2 | C3H5BrMg | 145.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1730-25-2
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00000044
InChI Key FEMBXICCJNZMMC-UHFFFAOYSA-M
Synonym allylmagnesium bromide, allyl magnesium bromide, allylmagnesium bromide solution, sjpdabpprujp@, allylmagnesium bromide solution, 1.0 m in diethyl ether, allylmagnesiumbromid, allylmagnesiumbromide, allyl magnesiumbromide, allyl-magnesium bromide, 2-propenylmagnesium bromide, grignard reagent
PubChem CID 74413
IUPAC Name magnesium;prop-1-ene;bromide
SMILES Br[Mg]CC=C

Specifications

CAS Min % 15.37, 81.22
CAS Max % 18.78, 84.64
Color Gray or Green-Yellow
Density 0.8510g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass bottle
Linear Formula H2C=CHCH2MgBr
Quantity 800 mL
UN Number 3399
Specific Gravity 0.851
Formula Weight 145.28
Physical Form Solution
Chemical Name or Material Allylmagnesium bromide, 1M solution in diethyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allylmagnesium bromide, 1M solution in diethylether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.