missing translation for 'onlineSavingsMsg'
Learn More

Allyltri-n-butyltin, 97%

Product Code. 11378955
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378955 25 g 25g
11368955 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378955 Supplier Thermo Scientific Alfa Aesar Supplier No. L14087.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24850-33-7 | C15H32Sn | 331.13 g/mol

Allyltri-n-butyltin acts as an allylation reagent for aldehydes catalyzed by chiral Lewis acids and ytterbium(III) trifluoromethanesulfonate. It is also used to prepare homoallylic alcohols with trichloro-1,3,5-triazene. Further, it plays an important role for tetrakis(triphenylphosphine)palladium(0) catalyzed coupling reaction with iodoquinones and allyl acetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltri-n-butyltin acts as an allylation reagent for aldehydes catalyzed by chiral Lewis acids and ytterbium(III) trifluoromethanesulfonate. It is also used to prepare homoallylic alcohols with trichloro-1,3,5-triazene. Further, it plays an important role for tetrakis(triphenylphosphine)palladium(0) catalyzed coupling reaction with iodoquinones and allyl acetates.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24850-33-7
Molecular Formula C15H32Sn
Molecular Weight (g/mol) 331.13
MDL Number MFCD00010346
InChI Key YLGRTLMDMVAFNI-UHFFFAOYSA-N
Synonym allyltributyltin, allyltributylstannane, allyltri-n-butyltin, stannane, tributyl-2-propenyl, tributyl prop-2-en-1-yl stannane, allyl tributyl stannane, unii-aor8od4103, tributyl-2-propenylstannane, tri-n-butyl-2-propenylstannane, allyl tributyltin
PubChem CID 90628
SMILES CCCC[Sn](CCCC)(CCCC)CC=C

Spezifikation

Density 1.079
Boiling Point 100°C to 102°C (0.5 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.486
Quantity 25 g
UN Number UN2788
Beilstein 3588340
Sensitivity Air sensitive
Solubility Information Immiscible with water.
IUPAC Name tributyl(prop-2-en-1-yl)stannane
Formula Weight 331.12
Percent Purity 97%
Chemical Name or Material Allyltri-n-butyltin
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.