missing translation for 'onlineSavingsMsg'
Learn More

Allyltrichlorosilane, 95%, Thermo Scientific™

Product Code. 10555111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10555111 25 g Glass bottle 25g
10061941 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10555111 Supplier Thermo Scientific Acros Supplier No. 313220250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-37-9
Molecular Formula C3H5Cl3Si
Molecular Weight (g/mol) 175.52
MDL Number MFCD00000486
InChI Key HKFSBKQQYCMCKO-UHFFFAOYSA-N
Synonym allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl
PubChem CID 7867
IUPAC Name trichloro(prop-2-enyl)silane
SMILES C=CC[Si](Cl)(Cl)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 117.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2CCHCH2SiCl3
Refractive Index 1.4430 to 1.4470
Quantity 25 g
Specific Gravity 1.2
Solubility Information Solubility in water: react violently with water
Formula Weight 175.52
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Allyltrichlorosilane, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.