missing translation for 'onlineSavingsMsg'
Learn More

Allyltrimethylsilane, 97%

Product Code. 10565491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10565491 50 mL Glass bottle 50mL
10215631 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10565491 Supplier Thermo Scientific Acros Supplier No. 349990500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 762-72-1 | C6H14Si | 114.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-72-1
Molecular Formula C6H14Si
Molecular Weight (g/mol) 114.27
MDL Number MFCD00008635
InChI Key HYWCXWRMUZYRPH-UHFFFAOYSA-N
Synonym allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane
PubChem CID 69808
IUPAC Name trimethyl(prop-2-enyl)silane
SMILES C[Si](C)(C)CC=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.7190g/mL
Boiling Point 85°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2C=CHCH2Si(CH3)3
Refractive Index 1.4060 to 1.408
Quantity 50 mL
Fieser 10,6; 11,16; 12,23; 13,11; 14,18; 15,8
Merck Index 15, 290
Specific Gravity 0.719
Solubility Information Solubility in water: insoluble
Formula Weight 114.27
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allyltrimethylsilane, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allyltrimethylsilane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.