missing translation for 'onlineSavingsMsg'
Learn More

Allyltrimethylsilane, TRC

Product Code. 30179397
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179397 10 g 10g
30178992 1 g 1g
30179147 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179397 Supplier Toronto Research Chemicals Supplier No. TRCA57235010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-72-1
Molecular Formula C6 H14 Si
Molecular Weight (g/mol) 114.26
Synonym Trimethyl-2-propen-1-yl-silane, Allyltrimethyl-silane, Trimethyl-2-propenyl-silane, 3-(Trimethylsilyl)-1-propene, 3-(Trimethylsilyl)propene, Trimethyl(2-propenyl)silane, Trimethylallylsilane
IUPAC Name trimethyl(prop-2-enyl)silane
SMILES C[Si](C)(C)CC=C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H14Si/c1-5-6-7(2,3)4/h5H,1,6H2,2-4H3
Formula Weight 114.0865
Chemical Name or Material Allyltrimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.