missing translation for 'onlineSavingsMsg'
Learn More

Almorexant, MedChemExpress

Product Code. 16476877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476877 5 mg 5mg
16435136 100 mg 100mg
16414178 10 mg 10mg
16410527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476877 Supplier Medchem Express Supplier No. HY10805
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Almorexant (ACT 078573) is an orally active, potent and competitive dual orexin receptor antagonist, with Kd values of 1.3 nM (OX1) and 0.17 nM (OX2), respectively. Almorexant reversibly blocks signaling of orexin-A and orexin-B peptides. Almorexant totally blocked the intracellular Ca2+ signal pathway. Almorexant stimulates caspase-3 activity in AsPC-1 cells and induces apoptosis.

Almorexant (ACT 078573) is an orally active, potent and competitive dual orexin receptor antagonist, with Kd values of 1.3 nM (OX1) and 0.17 nM (OX2), respectively. Almorexant reversibly blocks signaling of orexin-A and orexin-B peptides. Almorexant totally blocked the intracellular Ca2+ signal pathway. Almorexant stimulates caspase-3 activity in AsPC-1 cells and induces apoptosis.

Specifications

Chemical Name or Material Almorexant
CAS 871224-64-5
Color White
Purity Grade Notes Research
Molecular Formula C29H31F3N2O3
Quantity 5 mg
Synonym ACT 078573
Solubility Information DMSO : ≥ 46 mg/mL (89.75 mM)
SMILES CNC([C@@H](C1=CC=CC=C1)N2CCC3=CC(OC)=C(OC)C=C3[C@@H]2CCC4=CC=C(C(F)(F)F)C=C4)=O
Molecular Weight (g/mol) 512.56
Formula Weight 512.56
Percent Purity 99.01%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.