missing translation for 'onlineSavingsMsg'
Learn More

Almotriptan Malate, TRC

Product Code. 17998994
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17998994 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17998994 Supplier Toronto Research Chemicals Supplier No. TRCA57520010MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 181183-52-8
Molecular Formula C17 H25 N3 O2 S . C4 H6 O5
Quantity 10 mg
Synonym Butanedioic acid, 2-hydroxy-, compd. with N,N-dimethyl-5-[(1-pyrrolidinylsulfonyl)methyl]-1H-indole-3-ethanamine (1:1), Butanedioic acid, hydroxy-, compd. with 1-[[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]sulfonyl]pyrrolidine (1:1) (9CI), Pyrrolidine, 1-[[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]sulfonyl]-, hydroxybutanedioate (1:1) (9CI), Almotrip forte, Almotriptan malate, Axert, LAS 31416 D,L-malate acid, PNU 180638, PNU 180638E, LAS-31416 Malate;
InChI Formula InChI=1S/C17H25N3O2S.C4H6O5/c1-19(2)10-7-15-12-18-17-6-5-14(11-16(15)17)13-23(21,22)20-8-3-4-9-20;5-2(4(8)9)1-3(6)7/h5-6,11-12,18H,3-4,7-10,13H2,1-2H3;2,5H,1H2,(H,6,7)(H,8,9)
SMILES CN(C)CCc1c[nH]c2ccc(CS(=O)(=O)N3CCCC3)cc12.OC(CC(=O)O)C(=O)O
IUPAC Name N,N-dimethyl-2-[5-(pyrrolidin-1-ylsulfonylmethyl)-1H-indol-3-yl]ethanamine;2-hydroxybutanedioic acid
Molecular Weight (g/mol) 469.55
Formula Weight 469.1883
Chemical Name or Material Almotriptan Malate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.